missing translation for 'onlineSavingsMsg'
Learn More
Calcium sulfate dihydrate, Puriss., meets analytical spec. of NF, E 516, 99.0-101.0% (based on anhydrous substance), Honeywell Fluka™ Product Code.: 15641100

Calcium sulfate dihydrate, Puriss., meets analytical spec. of NF, E 516, 99.0-101.0% (based on anhydrous substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15641100 Shop All Honeywell Fluka Products
25 kg
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
This item is not returnable. View return policy

Product Code. 15641100

Brand: Honeywell-Fluka 1205625KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
IUPAC Name calcium;sulfate;dihydrate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

CAS Max % ≤100.0000%
Quantity 25 kg
Assay Percent Range 99.0 to 101.0%
Linear Formula CaSO4 · 2H2O
Grade puriss.
Packaging Paper / Poly Bag
Density 2.320 g/cm3
Chemical Name or Material Calcium sulfate dihydrate, Puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium sulfate dihydrate, Puriss., meets analytical spec. of NF, E 516, 99.0-101.0% (based on anhydrous substance), Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.