missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Hypochlorite, Technical, Fisher Chemical™
Chemical Identifiers
CAS | 7778-54-3 |
---|---|
Molecular Formula | CaCl2O2 |
Molecular Weight (g/mol) | 142.98 |
MDL Number | 10900 |
InChI Key | ZKQDCIXGCQPQNV-UHFFFAOYSA-N |
IUPAC Name | calcium dihypochlorite |
SMILES | [Ca++].[O-]Cl.[O-]Cl |
Chemical Identifiers
7778-54-3 | |
142.98 | |
ZKQDCIXGCQPQNV-UHFFFAOYSA-N | |
[Ca++].[O-]Cl.[O-]Cl |
CaCl2O2 | |
10900 | |
calcium dihypochlorite |
Specifications
100°C | |
White | |
Solid | |
10900 | |
[Ca++].[O-]Cl.[O-]Cl | |
142.98 | |
Plastic powder jar |
7778-54-3 | |
11.4 | |
CaCl2O2 | |
ZKQDCIXGCQPQNV-UHFFFAOYSA-N | |
calcium dihypochlorite | |
142.98g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Calcium hypochlorite
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H400-Very toxic to aquatic life.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Calcium Hypochlorite, Technical, Fisher Chemical™