missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride, 99%, Thermo Scientific Chemicals

Commercially important source for fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region.

£39.80 - £209.04

Chemical Identifiers

CAS 7789-75-5
Molecular Formula CaF2
Molecular Weight (g/mol) 78.08
MDL Number MFCD00010907
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
PubChem CID 84512
ChEBI CHEBI:35437
SMILES [F-].[F-].[Ca++]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11454293
View Documents
Thermo Scientific Alfa Aesar
A10913.22
100 g Glass bottle
£39.80
100g
Please to purchase this item. Need a web account? Register with us today!
11464293
View Documents
Thermo Scientific Alfa Aesar
A10913.36
500 g Plastic bottle
£124.62
500g
Please to purchase this item. Need a web account? Register with us today!
11474293
View Documents
Thermo Scientific Alfa Aesar
A10913.0E
2500 g Plastic bottle
£209.04
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calcium fluoride is the commercially important source for fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region. It is used in spectroscopy, thermal imaging systems and excimer lasers; it is also used in the manufacturing of optical components including windows, prisms and lenses. In addition, it is also used in aluminum-metallurgy, brake lining manufacturing industries, and in tooth enamel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-75-5
78.08
WUKWITHWXAAZEY-UHFFFAOYSA-L
84512
[F-].[F-].[Ca++]
CaF2
MFCD00010907
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
CHEBI:35437

Specifications

≈2500°C
7789-75-5
CaF2
EM>5,
Slightly soluble in acid. Insoluble in acetone.
[F-].[F-].[Ca++]
78.08
CHEBI:35437
99%
3.18
Calcium fluoride
1403°C
Crystals
MFCD00010907
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
WUKWITHWXAAZEY-UHFFFAOYSA-L
calcium;difluoride
84512
78.08
1.434
3.18
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 232-188-7

missing translation for 'rtecsNumber' : EW1760000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Zertifikate

RUO – Research Use Only