missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium carbonate, 97%, pure, chunks, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 kg
3 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 825.0°C |
Color | Beige-Gray to Beige-White |
Physical Form | Chunks |
Quantity | 3 kg |
Assay Percent Range | 96% min. (Complexometry) |
Linear Formula | CaCO3 |
Fieser | 01,103; 02,57; 04,67; 05,89 |
Merck Index | 15, 1659 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in diluted acids |
Formula Weight | 100.09 |
Show More |
Safety and Handling
Product Identifier
- Calcium carbonate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'