missing translation for 'onlineSavingsMsg'
Learn More

Calcium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals

£136.68 - £100460.00

Chemical Identifiers

CAS 71626-99-8
Molecular Formula Br2Ca
Molecular Weight (g/mol) 199.89
MDL Number MFCD00149608
InChI Key WGEFECGEFUFIQW-UHFFFAOYSA-L
IUPAC Name calcium dibromide
SMILES [Ca++].[Br-].[Br-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11415210
View Documents
Thermo Scientific Alfa Aesar
044497.14
25 g
£136.68
25g
Please to purchase this item. Need a web account? Register with us today!
11425210
View Documents
Thermo Scientific Alfa Aesar
044497.22
100 g
£404.01
100g
Please to purchase this item. Need a web account? Register with us today!
11435210
View Documents
Thermo Scientific Alfa Aesar
044497.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

71626-99-8
199.89
WGEFECGEFUFIQW-UHFFFAOYSA-L
[Ca++].[Br-].[Br-]
Br2Ca
MFCD00149608
calcium dibromide

Specifications

71626-99-8
Crystalline
MFCD00149608
[Ca++].[Br-].[Br-]
199.89
99.999%
Calcium bromide hydrate
White
Br2Ca
WGEFECGEFUFIQW-UHFFFAOYSA-L
calcium dibromide
199.90 (Anhydrous)
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P280-P305+P351+P338-P310

H318

missing translation for 'einecsNumber' : 232-164-6

missing translation for 'rtecsNumber' : EV9328000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only