missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf56 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25433-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody detects the endogenous protein (35 kDa) in mouse testis. The identity of an additional 65 kDa band is unknown.
C1orf56, also named as MENT, AF1Q and MLLT11, is involved in control of cellular proliferation. It is highly expressed in several cancers especially in neuroblastoma.Specifications
C1orf56 | |
Polyclonal | |
Unconjugated | |
MENT | |
AF1Q, C1orf56, MENT, MLLT11, RP11 316M1.9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
229588, 54964 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BUN1 | |
Gm128, MENT | |
C1orf56 Fusion Protein Ag13860 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |