missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf70 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20602-1-AP-150UL
This item is not returnable.
View return policy
Description
The MW of this protein is 48 kDa, and our antibody specifically recognizes the 48 kDa protein.
The function of C20orf30 has not been widely studied, and is yet to be fully elucidated.Specifications
C16orf70 | |
Polyclonal | |
Unconjugated | |
C16orf70 | |
C16orf6, C16orf70, lin 10, LIN10, UPF0183 protein C16orf70 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
102547323, 80262 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BSU1 | |
C16orf70, LOC102547323 | |
C16orf70 Fusion Protein Ag14576 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur