missing translation for 'onlineSavingsMsg'
Learn More

Butyl Acetate, Reagent Grade, 99.5%, Honeywell™

Product Code. 15628420
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Melting Point -78°C
Color Colorless
Density 0.88g/mL at 25°C
Boiling Point 126.2°C
Explosion Limit 0.076
Flash Point 23°C (73.4°F)
Ignition Point 790°F
Packaging Glass Bottle
Linear Formula CH3COO(CH2)3CH3
Refractive Index n20/D 1.394
Quantity 4 L
UN Number UN1123
Beilstein 1741921
Vapor Density 4 (vs air)
Formula Weight 116.16g/mol
Vapor Pressure 15mmHg (25°C), 8mmHg (20°C)
Percent Purity 99.50%
Chemical Name or Material Butyl acetate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'