Learn More
Fisherbrand™ Bulb for Diascopic Light
Illuminate Inverso TC100 Microscopes with this light bulb. Fisherbrand™ Bulbs for Diascopic Light are ergonomically designed for everyday use.
Brand: Fisherbrand™
Code : 04
Additional Details : Weight : 1.00000kg
Description
- G4 Halogen Bulb for diascopic light
- Accessory for microscopes: FB69155 and FB69156
- Available in packs of ten