missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 7.0 (20°C), Potassium Dihydrogen Phosphate / Disodium Hydrogen Phosphate, Traceable to SRM from NIST, with Fungicide, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
5 L
6 x 100 mL
6 x 500 mL
10 L
6 x 1 L
Description
Leading the Industry For Over 65 Years. Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured.
Composition per liter:
- 3.52 g Potassium dihydrogen phosphate (KH2PO4)
- 7.26 g di-Sodium hydrogen phosphate-2-hydrate (Na2HPO4*2H20)
Specifications
Specifications
Boiling Point | 100°C |
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000%, 0.7300%, 0.3500% |
CAS | 7558-79-4, 7778-77-0 |
Density | 1 g/cm3 |
Quantity | 6 x 1 L |
Solution Type | Buffer |
MDL Number | MFCD00146206 |
Packaging | Poly Bottle |
Chemical Name or Material | Buffer solution pH 7.0 (20°C), Potassium dihydrogen phosphate / disodium hydrogen phosphate, traceable to SRM from NIST, with fungicide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction