missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 1.00, Reagecon™
General use buffer solution. Reagecon™ Buffer Solution pH 1.00 has precision of ± 0.02 at 20°C.
£33.15 - £49.10
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11878783
|
Reagecon
10105 |
500 mL |
£33.15
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11460449
|
Reagecon
1010 |
1 L |
£49.10
1 L |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
General use buffer solution. Reagecon™ Buffer Solution pH 1.00 has precision of ± 0.02 at 20°C.
- General use buffer Solution
- pH 1.00 ± 0.02 at 20°C