missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 4.0 (20°C), With fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST, Honeywell Fluka™

Product Code. 15694020 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
5 L
10 L
6 x 1 L

With fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O

Specifications

pH 4.0
Quantity 1 L
UN Number NONH for all modes of transport
Chemical Name or Material Buffer solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.