missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Colour Coded Solution pH4.00 (Phthalate) Red, NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Brand: Fisher Chemical J/2825/21
Code : 23
Additional Details : Weight : 5.00000kg
Description
Ready to use for pH measurement.Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
- Standard and color-coded buffer solutions
- Manufactured to a tolerance of ±0.02 pH units
- Accurately standardized and labelled with lot-specific pH at 20°C
- All buffers are traceable to NIST and BS 1647 standard
- Packed in HDPE containers to help protect the products integrity an ensure safe handling (refers to 1L, 2.5L, and 5L quantities)
- Standard buffers are available over the range pH2 to pH10
- Color-coded buffer solutions are available over the range pH4, pH7, and pH
Specifications
Red | |
Liquid | |
217672 |
4 | |
5 L | |
Plastic container |