missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI Melting Point Monitor Software
For use with Melting Point apparatus.
Brand: BUCHI 11055332
Code : 61
Additional Details : Weight : 0.20000kg
Description
- For use with BUCHI Melting Point apparatus
- For observation of the melting curves and the melting video after measurement
- Scope of delivery: Software CD, USB cable and single PC license for Windows Vista and Windows XP
Specifications
Melting Point Monitor Software | |
Observation of the melting curves and the melting video after measurement | |
Melting Point apparatus |