Learn More
Hirschmann™ Class A Volumetric Flasks, Amber Glass
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks, Amber Glass are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2640266
 View more versions of this product
Product Code. 11682229
  • £67.80 / Pack of 2
Explore more special offers
Description
Description
  • With NS ground and hollow amber glass stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Specifications
DIN EN ISO 1042
Glass
20 mL
110 mm
12/21 NS Standard Ground
Yes
Brown
Brown
Borosilicate Glass, Duran Glass
±0.04 mL
±0.04 mL
A