missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Purple sodium salt, 0.04% w/v aq. soln., Thermo Scientific Chemicals
£24.32 - £37.99
Chemical Identifiers
CAS | 62625-30-3 |
---|---|
Molecular Formula | C21H15Br2NaO5S |
Molecular Weight (g/mol) | 562.204 |
MDL Number | MFCD00148896 |
InChI Key | KVSJYIXUWNOWBD-UGAWPWHASA-M |
Synonym | Bromocresol Purple, water soluble |
PubChem CID | 23665568 |
IUPAC Name | sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate |
SMILES | CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11368967
|
Thermo Scientific Alfa Aesar
038700.AE |
100 mL |
£24.32
100mL |
In Stock
Log in to see stock. |
|||||
11490151
|
Thermo Scientific Alfa Aesar
038700.AP |
500 mL |
£37.99
500mL |
In Stock
Log in to see stock. |
|||||
11378967
|
Thermo Scientific Alfa Aesar
038700.M0 |
6 x 100 mL | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62625-30-3 | |
562.204 | |
KVSJYIXUWNOWBD-UGAWPWHASA-M | |
23665568 | |
CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+] |
C21H15Br2NaO5S | |
MFCD00148896 | |
Bromocresol Purple, water soluble | |
sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate |
Specifications
62625-30-3 | |
C21H15Br2NaO5S | |
Bromocresol Purple, water soluble | |
CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+] | |
562.204 | |
562.22 | |
Liquid |
0.04% w/v aq. soln. | |
MFCD00148896 | |
KVSJYIXUWNOWBD-UGAWPWHASA-M | |
sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate | |
23665568 | |
Orange | |
Bromocresol Purple sodium salt |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title