missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Brefeldin A Solution (1000X)
Addition of Brefeldin A during the last hours of in vitro activation of cells results in enhanced detection of intracellular cytokines.
Brand: Invitrogen™ 00-4506-51
56.65 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Brefeldin A is an inhibitor of intracellular protein transport
- Incubation of cells in culture with Brefeldin A leads to blockade of protein transport to the Golgi complex (GC) and accumulation of proteins in the endoplasmic reticulum (ER)
- Brefeldin A is effective for enhanced detection of a majority of mouse and human intracellular cytokines; however, it is advised that the investigators evaluate the use and efficacy of this reagent as well as other protein transport inhibitors such as Monensin in their specific assay system
Specifications
1 mL | |
eBioscience™ | |
Liquid | |
Inhibitor |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- eBioscience Brefeldin A Solution (1000X)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methanol
For Research Use Only. Not for use in diagnostic procedures.