Learn More
Brady™ Vinyl Tapes (B-595) for BMP71 Printer
Indoor/Outdoor vinyl grade conforms to irregular, smooth or rough surfaces
Brand:  Brady™ 142384
Code : NEW
Additional Details : Weight : 0.14420kg
 View more versions of this product
Product Code. 11775603
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Vinyl film with permanent adhesive
  • Adheres well to pipes, walls, equipment, windows - most clean, dry surfaces
  • Resistant to grease, oil and most industrial chemicals
  • Five year outdoor durability in temperatures from 82° to -40°C (180° to -40°F, can be applied as low as -18°C (0°F)
Specifications
Specifications
Ochre
Vinyl Tape
BMP™71 Printer
50 ft. x 1 in.
50 ft.
1 in.
15.24 m
Vinyl (B-595)
Labeling pipes, walls, equipment, windows
1 Ea.
15.24 m x 2.54 cm (50 ft. x 1 in.)
15.24 m
2.54 cm
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats