missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Anti-Skid Floor Tapes

For indoor or outdoor use and floor marking needs

£87.85

Specifications

Adhesive Type Permanent Pressure Sensitive Acrylic
Material B-916 Grit-Coated Polyester
For Use With (Application) Floor marking needs
Length (English) 60 ft.
Length (Metric) 18.2 m
View More Specs

Products 1
Product Code Brand Color Width (Metric) Width (English) Price Quantity & Availability  
Product Code Brand Color Width (Metric) Width (English) Price Quantity & Availability  
10366574
View Documents
Brady™
078190
Black 5.08 cm 2 in.
£87.85
1 roll
In Stock
to see stock.
Add to basket
 
Description

Description

  • Made of rugged B-916 adhesive-backed polyester tape 
  • Coated with thousands of particles of durable aluminum oxide grit
  • Bonds permanently to clean, dry floors and withstands grease and oil
Specifications

Specifications

Permanent Pressure Sensitive Acrylic
Floor marking needs
18.2 m
B-916 Grit-Coated Polyester
60 ft.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.