Learn More
Trajan Scientific and Medical™ BP1 GC Capillary Columns
Suitable for all routine analysis. Trajan Scientific and Medical BP1 GC Capillary Columns are industry standard non polar phase columns.
Brand:  Trajan Scientific and Medical™ 054054
 View more versions of this product
Product Code. 15228633
  • £937.00 / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Specifications
0.22 mm
Hydrocarbon Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines
1 μm
100% Dimethyl Polysiloxane
BP1 GC Column
-60°C to +340/360°C
50 m
BP1