missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Borosilicate Glass Crude Filter Crucibles

Brand:  Robu™ 20602

Additional Details : Weight : 0.37810kg

 View more versions of this product

Product Code. 16803026

  • £226.00 / Pack of 10
Estimated Shipment: 21-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

34mm
For crude fiber extraction
34mm
Crude Filter Crucible
Borosilicate Glass
30 mL
60mm
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers