missing translation for 'onlineSavingsMsg'
Learn More

Fistreem International™ Boiling Flask

Suitable replacement boiling flask for the reliable and robust Loughborough Glass Water Still. Fistreem International™ Boiling Flask is constructed from robust borosilicate glass.

Brand:  Fistreem International™ W/FF9/8

Code : 11

Additional Details : Weight : 1.50000kg

Product Code. 13146000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Suitable for 8L/hr output Loughborough glass water still.
  • Constructed of robust borosilicate glass.
Specifications

Specifications

1.5 kg
8 L/hr. Output Loughborough Glass Still
Boiling Flask
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers