Learn More
Kautex™ Blue Polypropylene Screw Closures
Provides tamper evidence with pull-off ring and conical seal. Kautex™ Blue Polypropylene Screw Closures are ribbed for easy grip opening.
Brand:  Kautex™ 2000076305
 View more versions of this product
Product Code. 10289060
  • £1.75 / Each
Explore more special offers
Description
Description
  • Leakproof conical seal
  • Blue color for content identification
  • Polypropylene construction for strength and durability
  • Available for bottles with capacities of 10 to 2500 mL
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Blue
Polypropylene
2500 mL Bottle
Pull-off Ring and Conical Seal
Screw Cap
45 mm