Learn More
Tenak™ Blue Cardboard Cryobox
Cryobox made of water-repellent cardboard for long-lasting durability.
Brand: Tenak™ TE22082
Additional Details : Weight : 0.11000kg
Description
- Constructed of fibreboard/cardboard
- Water-repellent for longer durability
- Can be used with a large range of grid dividers
Specifications
Blue | |
Square | |
133 x 133 x 130 mm |
Cardboard | |
Cryo Box |