Learn More
OHAUS™ Block Cover
Click to view available options
Description:
Cover Low-Temperature 2 Block
Cover Low-Temperature 4 Block
Cover Low-Temperature 6 Block
Description
- Block heater accessories include modular blocks to accommodate almost any sample
- Modular blocks feature close contact of tubes-to-walls allowing for maximum heat retention
- Most blocks feature a thermometer well for measuring block temperature
- Stainless steel sand baths are available for irregular samples when there is no suitable block available
- Low temperature covers help to reduce the air flow for additional temperature stability
Specifications
Specifications
Description | Cover Low-Temperature 6 Block |
Type | Accessory, Block Cover |
For Use With (Application) | Low Temperatures |