missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Standard For ICP, 1000μg/mL in 10% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

£88.10 - £136.00

Chemical Identifiers

CAS 7440-69-9, 7697-37-2, 7732-18-5
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11408649
View Documents
SPEX Certiprep
PLBI4-2Y
125 mL Triple-leached LDPE bottle
£88.10
125mL
Please to purchase this item. Need a web account? Register with us today!
10090522
View Documents
SPEX Certiprep
PLBI4-2X
500 mL Triple-leached LDPE bottle
£136.00
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Bi1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7440-69-9, 7697-37-2, 7732-18-5
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
CHEBI:33301
[Bi]
Bi
MFCD00134033
5359367
bismuth

Specifications

7440-69-9, 7697-37-2, 7732-18-5
0.1, 10.0, 89.9
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
10% HNO3
AA, ICP
[Bi]
208.98
CHEBI:33301
Matrix Concentration Cat. No.
10% HNO3 1000μg/mL PLBI4-2Y, -2X
0.1, 10.0, 89.9
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Certified Reference Material
Bi
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth
5359367
Assurance
Bismuth Standard
Videos
Safety and Handling

Safety and Handling

missing translation for 'dotInformation' : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.