missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth Lead Tin Cadmium ingot (Wood's metal), Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
Wood's metal is useful as a low-melting solder, low-temperature casting metal, high temperature coupling fluid in heat baths, and as a fire-melted valve element in fire sprinkler systems in buildings. It is also used as a filler when bending thin-walled metal tube.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Gray |
Melting Point | 71°C |
Avg. Mol. Wt. or Mol. Wt. Range | Bi:Pb:Sn:Cd; 50:25:12.5:12.5 wt% |
UN Number | UN3077 |
Quantity | 100 g |
Solubility Information | Insoluble in water. |
Odor | Odorless |
Form | Ingot |
Assay | (Wood's metal) |
Chemical Name or Material | Bismuth Lead Tin Cadmium ingot (Wood's metal) |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Product Identifier
- Bismuth Lead Tin Cadmium ingot (Wood's metal)
- Danger
- Acute toxicity Category 4
- Acute toxicity Category 2
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1A
- Specific target organ toxicity Category 1
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H330-Fatal if inhaled.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H372-Causes damage to organs.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P201-Obtain special instructions before use.
- P320-Specific treatment is urgent (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Lead, Cadmium
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'