Learn More
Bismuth(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
£114.57 - £987.92
Chemical Identifiers
CAS | 1304-76-3 |
---|---|
Molecular Formula | Bi2O3 |
Molecular Weight (g/mol) | 465.96 |
MDL Number | MFCD00003462 |
InChI Key | TYIXMATWDRGMPF-UHFFFAOYSA-N |
IUPAC Name | dibismuth(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Bi+3].[Bi+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11380098
|
Thermo Scientific Alfa Aesar
042863.18 |
50 g |
£114.57
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11370098
|
Thermo Scientific Alfa Aesar
042863.30 |
250 g |
£359.79
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11390098
|
Thermo Scientific Alfa Aesar
042863.A1 |
1 kg |
£987.92
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bismuth(III) oxide is used in solid-oxide fuel cells. It is also used in optical glasses, to replace lead oxide in whitewares, luxes, varistor formulations and ceramic capacitor formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1304-76-3 | |
465.96 | |
TYIXMATWDRGMPF-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Bi+3].[Bi+3] |
Bi2O3 | |
MFCD00003462 | |
dibismuth(3+) trioxidandiide |
Specifications
∼1890°C | |
1304-76-3 | |
Powder | |
MFCD00003462 | |
Insoluble in water. Soluble in acids. | |
[O--].[O--].[O--].[Bi+3].[Bi+3] | |
465.96 | |
99.999% | |
8.9 g/mL | |
Bismuth (III) oxide |
817°C | |
Yellow | |
Bi2O3 | |
14,1273 | |
TYIXMATWDRGMPF-UHFFFAOYSA-N | |
dibismuth(3+) trioxidandiide | |
465.96 | |
Puratronic™ | |
(metals basis) |
Safety and Handling
missing translation for 'einecsNumber' : 215-134-7
missing translation for 'rtecsNumber' : EB2984460
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only