missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)carbodiimide, 98%, Thermo Scientific Chemicals

£22.20 - £206.00

Chemical Identifiers

CAS 1000-70-0
Molecular Formula C7H18N2Si2
Molecular Weight (g/mol) 186.4
MDL Number MFCD00051538
InChI Key KSVMTHKYDGMXFJ-UHFFFAOYSA-N
Synonym bis trimethylsilyl carbodiimide, carbodiimide, bis trimethylsilyl, n,n'-methanediylidenebis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, 1,3-bis trimethylsilyl carbodiimide, n,n'-bis trimethylsilyl carbodiimide, n,n'-methane tetraylbis 1,1,1-trimethylsilanamine, carbodiimide, bis trimethylsilyl-7ci,8ci, n,n'-methanetetraylbis 1,1,1-trimethylsilylamine
PubChem CID 70473
IUPAC Name N,N'-bis(trimethylsilyl)methanediimine
SMILES C[Si](C)(C)N=C=N[Si](C)(C)C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10006862
View Documents
Thermo Scientific Acros
375130010
1 g Glass bottle
£22.20
1g
Please to purchase this item. Need a web account? Register with us today!
10527323
View Documents
Thermo Scientific Acros
375130050
5 g Glass bottle
£51.30
5g
Please to purchase this item. Need a web account? Register with us today!
10736573
View Documents
Thermo Scientific Acros
375130250
25 g Glass bottle
£206.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Chemical Identifiers

1000-70-0
186.4
KSVMTHKYDGMXFJ-UHFFFAOYSA-N
70473
C[Si](C)(C)N=C=N[Si](C)(C)C
C7H18N2Si2
MFCD00051538
bis trimethylsilyl carbodiimide, carbodiimide, bis trimethylsilyl, n,n'-methanediylidenebis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, 1,3-bis trimethylsilyl carbodiimide, n,n'-bis trimethylsilyl carbodiimide, n,n'-methane tetraylbis 1,1,1-trimethylsilanamine, carbodiimide, bis trimethylsilyl-7ci,8ci, n,n'-methanetetraylbis 1,1,1-trimethylsilylamine
N,N'-bis(trimethylsilyl)methanediimine

Specifications

1000-70-0
Beige to Yellow or White
164°C
Authentic
C7H18N2Si2
MFCD00051538
bis trimethylsilyl carbodiimide, carbodiimide, bis trimethylsilyl, n,n'-methanediylidenebis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, 1,3-bis trimethylsilyl carbodiimide, n,n'-bis trimethylsilyl carbodiimide, n,n'-methane tetraylbis 1,1,1-trimethylsilanamine, carbodiimide, bis trimethylsilyl-7ci,8ci, n,n'-methanetetraylbis 1,1,1-trimethylsilylamine
KSVMTHKYDGMXFJ-UHFFFAOYSA-N
N,N'-bis(trimethylsilyl)methanediimine
70473
98%
Bis(trimethylsilyl)carbodiimide
-23°C
0.8210g/mL
42°C
97.5% min. (GC)
1.4330 to 1.435
0.821
Solubility in water: insoluble
C[Si](C)(C)N=C=N[Si](C)(C)C
186.4
186.4
Crystalline Powder, Crystals or Chunks
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 213-673-2

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only