missing translation for 'onlineSavingsMsg'
Learn More

Bis(tri-n-butyltin) sulfate, 97%, Thermo Scientific™

£15.17 - £57.36

Chemical Identifiers

CAS 26377-04-8
Molecular Formula C24H54O4SSn2
Molecular Weight (g/mol) 676.172
MDL Number MFCD00185908
InChI Key XEPUJTDDAPFGMG-UHFFFAOYSA-L
Synonym bis tri-n-butyltin sulfate, bis tributylstannylium sulfate, hexabutyl, i-sulphato 2--o,o:o',o' ditin
PubChem CID 15787681
IUPAC Name tributylstannanylium;sulfate
SMILES CCCC[Sn+](CCCC)CCCC.CCCC[Sn+](CCCC)CCCC.[O-]S(=O)(=O)[O-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11394567
View Documents
Thermo Scientific Alfa Aesar
039624.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11384567
View Documents
Thermo Scientific Alfa Aesar
039624.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

26377-04-8
676.172
XEPUJTDDAPFGMG-UHFFFAOYSA-L
15787681
CCCC[Sn+](CCCC)CCCC.CCCC[Sn+](CCCC)CCCC.[O-]S(=O)(=O)[O-]
C24H54O4SSn2
MFCD00185908
bis tri-n-butyltin sulfate, bis tributylstannylium sulfate, hexabutyl, i-sulphato 2--o,o:o',o' ditin
tributylstannanylium;sulfate

Specifications

26377-04-8
MFCD00185908
bis tri-n-butyltin sulfate, bis tributylstannylium sulfate, hexabutyl, i-sulphato 2--o,o:o',o' ditin
CCCC[Sn+](CCCC)CCCC.CCCC[Sn+](CCCC)CCCC.[O-]S(=O)(=O)[O-]
676.172
676.14
Powder
C24H54O4SSn2
UN3146
XEPUJTDDAPFGMG-UHFFFAOYSA-L
tributylstannanylium;sulfate
15787681
97%
Bis(tri-n-butyltin) sulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

missing translation for 'einecsNumber' : 247-634-6

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications