missing translation for 'onlineSavingsMsg'
Learn More

Bis(carboxyethylgermanium) sesquioxide, 99.7%, Thermo Scientific Chemicals

£48.24 - £549.74

Chemical Identifiers

CAS 12758-40-6
Molecular Formula C6H10Ge2O7
Molecular Weight (g/mol) 339.399
MDL Number MFCD00049319
InChI Key XEABSBMNTNXEJM-UHFFFAOYSA-N
Synonym propagermanium, proxigermanium, carboxyethylgermanium sesquioxide, 14c-proxigermanium, 2-carboxyethylgermasesquioxane, dipropanoic acid germanium sequioxide, 2-carboxyethyl germanium sesquioxide, bis 2-carboxyethylgermanium sesquioxide, ccris 5469, carboxylethylgermanium sesquioxide
PubChem CID 83030
ChEBI CHEBI:32060
IUPAC Name 3-[[2-carboxyethyl(oxo)germyl]oxy-oxogermyl]propanoic acid
SMILES C(C[Ge](=O)O[Ge](=O)CCC(=O)O)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11314277
SDS View Documents
Thermo Scientific Alfa Aesar
036758.06
5 g
£48.24
5g
Please to purchase this item. Need a web account? Register with us today!
11324277
SDS View Documents
Thermo Scientific Alfa Aesar
036758.14
25 g
£158.79
25g
Please to purchase this item. Need a web account? Register with us today!
11304277
SDS View Documents
Thermo Scientific Alfa Aesar
036758.22
100 g
£549.74
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A bifunctional metalloligand with metal-C (Ge-C) bonds,bis(carboxyethylgermanium) sesquioxide, has been used to make two metal-organometallic frameworks (MOMFs). bis-(carboxyethylgermanium)sesquioxide is a as a promising anti-oxidative agent. Bis(carboxyethylgermanium) sesquioxide induce INF-γ and has undergone clinical trials with promising preclinical results.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12758-40-6
339.399
XEABSBMNTNXEJM-UHFFFAOYSA-N
83030
3-[[2-carboxyethyl(oxo)germyl]oxy-oxogermyl]propanoic acid
C6H10Ge2O7
MFCD00049319
propagermanium, proxigermanium, carboxyethylgermanium sesquioxide, 14c-proxigermanium, 2-carboxyethylgermasesquioxane, dipropanoic acid germanium sequioxide, 2-carboxyethyl germanium sesquioxide, bis 2-carboxyethylgermanium sesquioxide, ccris 5469, carboxylethylgermanium sesquioxide
CHEBI:32060
C(C[Ge](=O)O[Ge](=O)CCC(=O)O)C(=O)O

Specifications

12758-40-6
Odorless
MFCD00049319
Slightly soluble in water.
C(C[Ge](=O)O[Ge](=O)CCC(=O)O)C(=O)O
339.399
CHEBI:32060
99.7%
Bis(carboxyethylgermanium) sesquioxide
320°C (decomposition)
C6H10Ge2O7
propagermanium, proxigermanium, carboxyethylgermanium sesquioxide, 14c-proxigermanium, 2-carboxyethylgermasesquioxane, dipropanoic acid germanium sequioxide, 2-carboxyethyl germanium sesquioxide, bis 2-carboxyethylgermanium sesquioxide, ccris 5469, carboxylethylgermanium sesquioxide
XEABSBMNTNXEJM-UHFFFAOYSA-N
3-[[2-carboxyethyl(oxo)germyl]oxy-oxogermyl]propanoic acid
83030
339.25
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 235-800-0

missing translation for 'rtecsNumber' : UA2459750

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.