missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-3-boronic acid, 98%, Thermo Scientific Chemicals

£30.90

Chemical Identifiers

CAS 5122-95-2
Molecular Formula C12H11BO2
Molecular Weight (g/mol) 198.028
MDL Number MFCD01318102
InChI Key GOXICVKOZJFRMB-UHFFFAOYSA-N
Synonym 3-biphenylboronic acid, biphenyl-3-boronic acid, 1,1'-biphenyl-3-ylboronic acid, biphenyl-3-ylboronic acid, 3-phenylphenyl boranediol, 3-phenylphenyl boronic acid, 3-biphenyl boronic acid, 3-phenylbenzeneboronic acid, 3-biphenylylboronic acid, 1,1'-biphenyl-3-yl boronic acid
PubChem CID 2734315
IUPAC Name (3-phenylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)C2=CC=CC=C2)(O)O
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11368496
View Documents
Thermo Scientific Alfa Aesar
L17552.03
1 g
£30.90
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is a biological reagent used as a boronate-assisted fluorogenic chemosensor. It is evaluated for pharmacological activity as fatty acid amide hydrolase inhibitors. It is a reactant involved in coupling with potassium cyanate, quinones, or fluorous tagged N-hydroxyphthalimide and direct C-H arylation of electron-deficient heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a biological reagent used as a boronate-assisted fluorogenic chemosensor. It is evaluated for pharmacological activity as fatty acid amide hydrolase inhibitors. It is a reactant involved in coupling with potassium cyanate, quinones, or fluorous tagged N-hydroxyphthalimide and direct C-H arylation of electron-deficient heterocycles.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

5122-95-2
198.028
GOXICVKOZJFRMB-UHFFFAOYSA-N
2734315
B(C1=CC(=CC=C1)C2=CC=CC=C2)(O)O
C12H11BO2
MFCD01318102
3-biphenylboronic acid, biphenyl-3-boronic acid, 1,1'-biphenyl-3-ylboronic acid, biphenyl-3-ylboronic acid, 3-phenylphenyl boranediol, 3-phenylphenyl boronic acid, 3-biphenyl boronic acid, 3-phenylbenzeneboronic acid, 3-biphenylylboronic acid, 1,1'-biphenyl-3-yl boronic acid
(3-phenylphenyl)boronic acid

Specifications

5122-95-2
Odorless
MFCD01318102
3-biphenylboronic acid, biphenyl-3-boronic acid, 1,1'-biphenyl-3-ylboronic acid, biphenyl-3-ylboronic acid, 3-phenylphenyl boranediol, 3-phenylphenyl boronic acid, 3-biphenyl boronic acid, 3-phenylbenzeneboronic acid, 3-biphenylylboronic acid, 1,1'-biphenyl-3-yl boronic acid
GOXICVKOZJFRMB-UHFFFAOYSA-N
(3-phenylphenyl)boronic acid
2734315
98%
206°C to 211°C
C12H11BO2
2836311
Insoluble in water.
B(C1=CC(=CC=C1)C2=CC=CC=C2)(O)O
198.028
198.03
Biphenyl-3-boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only