missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ Phosphate Buffered Solution (1X)
Serves as a wash solution in a wide variety of laboratory applications. BioWhittaker™ Phosphate Buffered Solution (1X) Phosphate Buffered Saline is manufactured in accordance with cGMP regulations.
£18.59
Specifications
Additive | No Calcium, No Magnesium |
---|---|
Content And Storage | 15° to 30°C |
Product Type | Phosphate Buffered Saline (PBS) |
pH | 7.3 to 7.5 |
Sterility | Sterile |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11629980
|
Lonza™
BE17-516F |
500 mL | N/A | ||||||
Description
- Wash solution
- Manufactured in accordance with cGMP regulations
- Cell culture tested to assure compatibility
Specifications
No Calcium, No Magnesium | |
Phosphate Buffered Saline (PBS) | |
Sterile |
15° to 30°C | |
7.3 to 7.5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title