Learn More
Agilent BioTek MultiFlo FX Microplate Dispenser Accessory, Reagent Bottle
Provides supply containment
Brand: Agilent BioTek 7140511
Additional Details : Weight : 0.90000kg
Description
- Autoclavable
- With cap and connector
Specifications
Reagent Bottle | |
CE and TUV marked. RoHS compliant. | |
Cap, connector |
Autoclavable | |
For microfill, multiflo fx, precision, bio-tek dispensers and pipetting systems with autoclavable fluid paths |