missing translation for 'onlineSavingsMsg'
Learn More

BioAcryl-P (40%, 19:1), Thermo Scientific Chemicals

BioAcryl-P (Acrylamide/Bisacrylamide 29:1, CAS # 25034-58-6) is a powder-based formula used to create the gel matrix necessary for electrophoresis experiments.

Brand:  Thermo Scientific Alfa Aesar J60909.K2


 View more versions of this product

Product Code. 15433599

  • £169.85 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Acrylamide/Bisacrylamide premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• BioAcryl-P is a 29:1 ratio of Acrylamide/Bisacrylamide
• Acrylamide is a crystalline amide that forms polymer structures at room temperature
• Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation
Application
• Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Acrylamide/Bisacrylamide 19:1
40% soln
1 L
UN3426
Liquid
Colorless
MFCD00080848
It is soluble in water.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BioAcryl-P (40%, 19:1)

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
  • H372-Causes damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Acrylamide, Methylene diacrylamide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only