missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BICINE, 0.5M buffer soln., pH 8.5

£67.64 - £106.53

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15424199
View Documents
Thermo Scientific Chemicals
J61632.AE
100 mL
£67.64
100mL
Please to purchase this item. Need a web account? Register with us today!
15434199
View Documents
Thermo Scientific Chemicals
J61632.AK
250 mL
£106.53
250mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A zwitterionic amino acid buffer agent used for low temperature work. The use of bicine in a thin layer ion exchange chromatography method for protein resolution has been published. Bicine has been utilized in peptide and protein crystallization. A kinetic study of a quaternary transition-state analogue complex of creatine kinase used bicine in the reaction buffer. A multiphasic buffer system for SDS-PAGE of proteins and peptides that incorporates bicine has been described.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A zwitterionic amino acid buffer agent used for low temperature work. The use of bicine in a thin layer ion exchange chromatography method for protein resolution has been published. Bicine has been utilized in peptide and protein crystallization. A kinetic study of a quaternary transition-state analogue complex of creatine kinase used bicine in the reaction buffer. A multiphasic buffer system for SDS-PAGE of proteins and peptides that incorporates bicine has been described.

Solubility
Soluble in water (1M) at 20°C, ethanol (slightly), ether (slightly), and chloroform (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

150-25-4
185.16
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
CHEBI:40957
[Na+].OCCN(CCO)CC([O-])=O
C6H12NNaO4
MFCD00004295
8761
2-[bis(2-hydroxyethyl)amino]acetic acid

Specifications

150-25-4
8.5
C6H12NNaO4
Soluble in water (1M) at 20°C,ethanol (slightly),ether (slightly),and chloroform (slightly).
[Na+].OCCN(CCO)CC([O-])=O
185.16
CHEBI:40957
0.5 M
Liquid
MFCD00004295
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
2-[bis(2-hydroxyethyl)amino]acetic acid
8761
BICINE buffer soln.
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only