missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl alcohol, Puriss. p.a., ACS reagent, ≥99.0% (GC), Honeywell Riedel-de Haën™
Puriss. p.a., ACS reagent,≥99.0% (GC)
£78.65 - £266.95
Chemical Identifiers
CAS | 100-51-6 |
---|---|
Molecular Formula | C7H8O |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00004599,MFCD03792087 |
InChI Key | WVDDGKGOMKODPV-UHFFFAOYSA-N |
Synonym | benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol |
PubChem CID | 244 |
ChEBI | CHEBI:17987 |
IUPAC Name | phenylmethanol |
SMILES | OCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15631270
|
Honeywell-Riedel-de Haen
13160-4X25ML |
4 x 25 mL | Glass Bottle | N/A | |||||
15641270
|
Honeywell-Riedel-de Haen
13160-500ML |
500 mL | Glass Bottle |
£78.65
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15621270
|
Honeywell-Riedel-de Haen
13160-2.5L |
2.5 L | Glass Bottle |
£266.95
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
100-51-6 | |
108.14 | |
WVDDGKGOMKODPV-UHFFFAOYSA-N | |
244 | |
phenylmethanol |
C7H8O | |
MFCD00004599,MFCD03792087 | |
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
CHEBI:17987 | |
OCC1=CC=CC=C1 |
Specifications
100-51-6 | |
-16°C to -13°C (literature) | |
205.3°C | |
C7H8O | |
MFCD00004599,MFCD03792087 | |
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
OCC1=CC=CC=C1 | |
108.14 | |
CHEBI:17987 | |
>99% | |
Benzyl alcohol, Puriss. p.a., ACS reagent, ≥99.0% (GC) |
≤100.0000% | |
1.045 g/mL at 25 °C(lit.) | |
100.4°C | |
C6H5CH2OH | |
878307 | |
WVDDGKGOMKODPV-UHFFFAOYSA-N | |
phenylmethanol | |
244 | |
108.14 | |
ACS Reagent |
Safety and Handling
missing translation for 'shelfLife' : 360 days from date of manufacture
missing translation for 'dotInformation' : Packing Group: 3
missing translation for 'einecsNumber' : 202-859-9
Recommended Storage : Room Temp
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title