missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Wire Rack ID Tags
Eliminate the challenges of labeling wire racks, trays, baskets and other wire-formed items
Brand: Bel-Art™ F13446-0000
Code : 03
Additional Details : Weight : 1.00000kg
Description
- Durable, reusable and easy to snap on or pull off
- Smooth, polypropylene surface allows labeling directly onto the tag with permanent marker, dry-erase marker or a pressure-sensitive adhesive label
- Fits wire-formed items made of 0.125 in. (3mm) or 0.188 in. (5mm) wire
- Chemical resistant and autoclavable at 121°C (250°F)
- W × H × D: 3.5 × 1.5 × 0.125 in. (8.9 × 3.8 × 0.3mm)
Specifications
1.5 in. | |
3.81cm | |
3.5 in. | |
8.89 cm |