missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Standards Bottle Rack
Organizes valuable USP™ reference standards bottles for convenient storage and access
Brand: Bel-Art™ F18513-2300
80.41 GBP valid until 2024-04-26
Use promo code "21699" to get your promotional price.
Code : A0
Additional Details : Weight : 0.19800kg
Description
- Suitable for use in Secador™ 1.0 to 4.0 desiccators for low humidity storage
- Clear acrylic construction for easy viewing of labels
- Stores thirty 23mm standards bottles in a compact footprint
Specifications
Bottle Rack | |
Clear | |
30 | |
33 cm | |
11 cm | |
2.2 cm |
Not Autoclavable | |
Acrylic | |
13 x 3.37 x 0.87 in. (33 x 11 x 2.2cm) | |
13 in. | |
3.37 in. | |
0.87 in. |