Learn More
Bel-Art™ Delrin™ Spring Tubing Clamps
Self-closing spring operated tubing clamp is suitable for all flexible tubing
Brand: Bel-Art™ H18213-0000
This item is not returnable.
View return policy
Description
- Positive-actionclamp opens by squeezing handles together
- Naturally clamps shut to stop flow
- Made from chemical- and stain-resistant Delrin™ plastic and stainless steel
Specifications
3 | |
Acetal Plastic | |
White | |
8 mm |
Spring Style Tubing Clamp | |
All flexible .31 in. (.78cm) O.D. tubing | |
Angled |