missing translation for 'onlineSavingsMsg'
Learn More

Beaker Bristle Brushes

Features galvanized steel wire handle. Pinnacle Brush™ Beaker Bristle Brushes offer gentle cleaning for beakers.

Brand:  PINNACLE Brush D1225/2

Code : 03

Additional Details : Weight : 0.01350kg

Product Code. 12358328

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Wire handle
Specifications

Specifications

Bottle Brush
445 mm
80 x 28 mm
Steel Wire
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers