Learn More
BD Mouse TNF (Mono/Mono) ELISA Set
Determination of mouse tumor necrosis factor
Brand:  BD 555268
827.67 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15818038
  • £839.00 / 1 set
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
Alert:
Toxic if ingested, inhaled, or in contact with skin
Includes: Capture Antibody: Anti-mouse TNF-α , Detection Antibody: Biotinylated anti-mouse TNF-α, Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate, Standard: Recombinant mouse TNF-α (lyophilized)
Requires : 96-well BD Falcon™ ELISA plates (Cat. No. 353279) are recommended, microplate reader capable of measuring absorbance at 450nm, precision pipettes, graduated cylinder, deionized or distilled water, wash bottle or automated washer, log-log graph paper or automated data reduction, tubes to prepare standard dilutions, laboratory timer, plate sealers or parafilm
Description
Description
  • For natural or recombinant mouse TNF in serum, plasma, and cell culture supernatants
  • Allow flexible assay design
  • Calibrated against purified Baculovirus-expressed recombinant mouse TNF
  • Intended for use as integral unit
ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
615.6 to 1000pg/mL
20 96-well Plates
anti-Mouse TNF monoclonal (capture) antibody, Biotinylated anti-Mouse TNF monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse TNF (standard), Streptavidin-HRP Conjugate
2°C to 8°C
Mouse TNF (Mono/Mono) ELISA set
To develop ELISA for natural or recombinant Mouse TNF in Serum, Plasma and cell culture supernatants
20 Plates
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research use only