Learn More
BD Mouse IL-5 ELISA Set
An immunoassay with different sensitivity and dynamic range
Brand:  BD 555236
798.48 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15848028
  • £808.00 / 1 set
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
Includes: capture antibody, detection antibody, enzyme reagent, standards, instruction/analysis certificate
Description
Description
To develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant mouse IL-5 in serum, plasma, and cell culture supernatants.
Enzyme-linked immunosorbent assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
15.6 to 1000pg/mL
20 96-well Plates
anti-Mouse IL-5 monoclonal (capture) antibody, Biotinylated anti-Mouse IL-5 monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse IL-5 (standard), Streptavidin-HRP Conjugate
2°C to 8°C
Mouse IL-5 ELISA set
To develop ELISA for natural or recombinant Mouse IL-5 in Serum, Plasma and cell culture supernatants
20 Plates
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research Use Only