missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Human IL-6 ELISA Set
An immunoassay with different sensitivity and dynamic range
Brand: BD 555220
862.51 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: Capture antibody, detection antibody, enzyme reagent, standards, instruction/analysis certificate
Description
Used to develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant human IL-6 in serum, plasma, and cell culture supernatants.
Enzyme-linked immunosorbent assay
Specifications
4.7 to 300pg/mL | |
Twenty 96-well Plates | |
anti-Human IL-6 monoclonal (capture) Antibody, Biotinylated anti-Human IL-6 monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human IL-6 (standard), Streptavidin-HRP Conjugate |
Human IL-6 ELISA Set | |
To develop ELISA for natural or recombinant Human IL-6 in serum, plasma and cell culture supernatants | |
2°C to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction