missing translation for 'onlineSavingsMsg'
Learn More

BAX Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen BAX
Concentration 340 μg/mL
Applications Western Blot, Immunoprecipitation, Immunohistochemistry, Flow Cytometry, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16806255
View Documents Promotion Details
Proteintech
50599-2-IG-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16896245
View Documents Promotion Details
Proteintech
50599-2-IG-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The BAX antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BAX. This antibody recognizes human, mouse, rat antigen. The BAX antibody has been validated for the following applications: FC, IP, WB, ELISA analysis.

Specifications

Specifications

BAX
Western Blot, Immunoprecipitation, Immunohistochemistry, Flow Cytometry, ELISA
Unconjugated
Human, Rat, Mouse
IgG
340 μg/mL
Polyclonal
Rabbit
581
Primary
Videos
SDS
Documents

Documents

Product Certifications