Learn More
Memmert™ Battery-Buffered ControlCOCKPIT
Uninterrupted supply for the entire display unit (ControlCOCKPIT)
Brand:  Memmert™ ICO105C2
 View more versions of this product
Product Code. 17199914
  • £851.00 / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Complete documentation of all parameters even when there is a power failure
  • The CO2 parameter is continuously regulated
Specifications
Specifications
ICO105
ControlCOCKPIT