missing translation for 'onlineSavingsMsg'
Learn More
Barium nitrate, Puriss. p.a., ACS Reagent, ≥99%, ≥52% Ba basis, Honeywell Fluka™ Product Code.: 15663260

Barium nitrate, Puriss. p.a., ACS Reagent, ≥99%, ≥52% Ba basis, Honeywell Fluka™

Product Code. 15663260 Shop All Honeywell Fluka Products
500 g
Click to view available options
Quantity:
500 g
5 kg
6 x 500 g
25 kg
This item is not returnable. View return policy

Product Code. 15663260

Brand: Honeywell-Fluka 31128500G

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99%,≥52% Ba basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2Show MoreShow Less
PubChem CID 24798
IUPAC Name barium(2+);dinitrate

Specifications

Quantity 500 g
Linear Formula Ba(NO3)2
UN Number 1446
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Formula Weight 261.34g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Packaging Plastic Bottle
Chemical Name or Material Barium Nitrate, ACS reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium nitrate, Puriss. p.a., ACS Reagent, ≥99%, ≥52% Ba basis, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.