missing translation for 'onlineSavingsMsg'
Learn More
Barium hydroxide octahydrate, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥98%, Honeywell Fluka™ Product Code.: 15633260

Barium hydroxide octahydrate, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥98%, Honeywell Fluka™

Product Code. 15633260 Shop All Honeywell Fluka Products
500 g
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 15633260

Brand: Honeywell-Fluka 31127500G

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrateShow MoreShow Less
PubChem CID 17749109
IUPAC Name barium(2+);dihydroxide;octahydrate
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

Quantity 500 g
Linear Formula Ba(OH)2 · 8H2O
UN Number 3262
Formula Weight 315.46g/mol
Percent Purity ≥98%
Grade puriss. p.a.
Packaging Plastic Bottle
Chemical Name or Material Barium Hydroxide Octahydrate, Reag. Ph. Eur. reag. ISO
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium hydroxide octahydrate, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥98%, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.