missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Bacto™ Neopeptone
An animal origin (AO) enzymatic digest of bovine (milk) and porcine protein. This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation.
£6790.00
Specifications
Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
---|---|
Form | Powder |
For Use With (Application) | Production of vaccines for animal health applications |
Product Line | Bacto™ |
Source | Animal origin, bovine and porcine |
Description
- Bacto Neopeptone is an animal origin (AO) enzymatic digest of bovine (milk) and porcine protein
- This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation
- It has been used as a component of culture media in the cultivation of human pathogens, notably Bordetella pertussis and group A streptococci
- Also used with Borrelia burgdorferi and Clostridium species, to produce vaccines for animal health application
- Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Applications of Bacto Neopeptone include:
- Recommended for use in media for detection of fungi
- Reported to provide nutrients for support of spirochetes and protozoa
- Microbial fermentation
- Production of vaccines for animal health applications
Specifications
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. | |
Production of vaccines for animal health applications | |
Animal origin, bovine and porcine |
Powder | |
Bacto™ |