missing translation for 'onlineSavingsMsg'
Learn More

Azure A, Thermo Scientific Chemicals

For staining cell granules and tissues

£25.53 - £55.17

Chemical Identifiers

CAS 531-53-3
Molecular Formula C14H14ClN3S
Molecular Weight (g/mol) 291.797
MDL Number MFCD00012112
InChI Key NALREUIWICQLPS-UHFFFAOYSA-N
Synonym C.I. 52005
PubChem CID 13735
IUPAC Name (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
SMILES C[N+](=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1)C.[Cl-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15453949
SDS View Documents
Thermo Scientific Alfa Aesar
J61346.14
25 g
£25.53
25g
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
 
15463949
SDS View Documents
Thermo Scientific Alfa Aesar
J61346.22
100 g
£55.17
100g
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
Description

Description

For staining cell granules and tissues

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

531-53-3
291.797
NALREUIWICQLPS-UHFFFAOYSA-N
13735
C[N+](=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1)C.[Cl-]
C14H14ClN3S
MFCD00012112
C.I. 52005
(7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride

Specifications

290°C (decomposition)
C14H14ClN3S
3922287
C.I. 52005
C[N+](=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1)C.[Cl-]
291.797
291.8
Powder
531-53-3
MFCD00012112
14,927
NALREUIWICQLPS-UHFFFAOYSA-N
(7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
13735
Green
Azure A
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 208-510-7

missing translation for 'rtecsNumber' : SP5660000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.